missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyanoacetyl)pyrrolidine, 98+%

Catalog No. AAA1252303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1252303 1 g
AAA1252306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1252303 Supplier Thermo Scientific Chemicals Supplier No. A1252303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14227-95-3 | C7H10N2O | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14227-95-3
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00020838
InChI Key VEUDVNNBYYRZBV-UHFFFAOYSA-N
Synonym 1-cyanoacetyl pyrrolidine, 3-oxo-3-pyrrolidin-1-yl propanenitrile, 1-cyanoacetylpyrrolidine, 3-oxo-3-1-pyrrolidinyl propanenitrile, pyrrolidine, 1-cyanoacetyl, 1-pyrrolidinepropanenitrile,b-oxo, 3-oxo-3-pyrrolidinylpropanenitrile, n-cyanoacetylpyrrolidine, acmc-20ak5f, oxopyrrolidinylpropanenitrile
PubChem CID 84272
IUPAC Name 3-oxo-3-pyrrolidin-1-ylpropanenitrile
SMILES O=C(CC#N)N1CCCC1

Specifications

Melting Point 71°C to 74°C
Odor Odorless
Quantity 1 g
Beilstein 637151
Formula Weight 138.17
Percent Purity ≥98%
Chemical Name or Material 1-(Cyanoacetyl)pyrrolidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 238-096-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only