missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%

Catalog No. AAL0836914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0836914 25 g
AAL0836906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0836914 Supplier Thermo Scientific Chemicals Supplier No. L0836914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36140-83-7 | C8H9N3O | 163.18 g/mol

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.

Chemical Identifiers

CAS 36140-83-7
Molecular Formula C8H9N3O
Molecular Weight (g/mol) 163.18
MDL Number MFCD00051954
InChI Key DDWZYWSLHBDVGR-UHFFFAOYSA-N
Synonym 1-cyanoacetyl-3,5-dimethylpyrazole, 3-3,5-dimethyl-1h-pyrazol-1-yl-3-oxopropanenitrile, 3-3,5-dimethylpyrazol-1-yl-3-oxopropanenitrile, 1-cyanoacetyl-3,5-dimethyl-1h-pyrazole, 3-3,5-dimethylpyrazolyl-3-oxopropanenitrile, cambridge id 5140887, 1h-pyrazole-1-propanenitrile, 3,5-dimethyl-, a-oxo, 1,cyanoacetyl-3,5-dimethylpyrazole, 1h-pyrazole-1-propanenitrile, 3,5-dimethyl-beta-oxo
PubChem CID 262101
IUPAC Name 3-(3,5-dimethylpyrazol-1-yl)-3-oxopropanenitrile
SMILES CC1=CC(=NN1C(=O)CC#N)C

Specifications

Melting Point 118°C to 122°C
Quantity 25 g
Beilstein 6286
Sensitivity Hygroscopic
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only