CAS RN 36140-83-7
IUPAC Name:
3-(3,5-dimethyl-1H-pyrazol-1-yl)-3-oxopropanenitrile
Synonyms:
3-(3,5-dimethylpyrazol-1-yl)-3-oxopropanenitrile
Molecular Weight (g/mol):
163.18
Molecular Formula:
C8H9N3O
InChi Key:
DDWZYWSLHBDVGR-UHFFFAOYSA-N
SMILES:
CC1=NN(C(=O)CC#N)C(C)=C1
1
–
1
of
1
results
1
Promotions Available
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%
CAS: 36140-83-7 Molecular Formula: C8H9N3O Molecular Weight (g/mol): 163.18 MDL Number: MFCD00051954 InChI Key: DDWZYWSLHBDVGR-UHFFFAOYSA-N Synonym: 1-cyanoacetyl-3,5-dimethylpyrazole,3-3,5-dimethyl-1h-pyrazol-1-yl-3-oxopropanenitrile,3-3,5-dimethylpyrazol-1-yl-3-oxopropanenitrile,1-cyanoacetyl-3,5-dimethyl-1h-pyrazole,3-3,5-dimethylpyrazolyl-3-oxopropanenitrile,cambridge id 5140887,1h-pyrazole-1-propanenitrile, 3,5-dimethyl-,a-oxo,1,cyanoacetyl-3,5-dimethylpyrazole,1h-pyrazole-1-propanenitrile, 3,5-dimethyl-beta-oxo PubChem CID: 262101 IUPAC Name: 3-(3,5-dimethylpyrazol-1-yl)-3-oxopropanenitrile SMILES: CC1=CC(=NN1C(=O)CC#N)C