missing translation for 'onlineSavingsMsg'
Learn More

1-Cyano-1-cyclopropanecarboxylic acid, 97%

Catalog No. aah2773003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2773003 1 g
AAH2773006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2773003 Supplier Thermo Scientific Chemicals Supplier No. H2773003
Only null left

CAS: 6914-79-0 | C5H4NO2 | 110.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6914-79-0
Molecular Formula C5H4NO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00190651
InChI Key KSJJMSKNZVXAND-UHFFFAOYSA-M
Synonym 1-cyanocyclopropanecarboxylic acid, 1-cyano-1-cyclopropanecarboxylic acid, cyanocyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-cyano, cyclopropane, 4, 1-cyanocyclopropane-carboxylic acid, 1-cyano-1-cyclopropanecarboxylicacid, 1-cyano-cyclo-propanecarboxylic acid, 1-cyano-1-cyclopropane carboxylic acid, 1-cyano-1-cyclopropane-carboxylic acid
PubChem CID 2733259
IUPAC Name 1-cyanocyclopropane-1-carboxylic acid
SMILES [O-]C(=O)C1(CC1)C#N

Specifications

Melting Point 143°C to 149°C
Quantity 1 g
UN Number UN2923
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 1-Cyano-1-cyclopropanecarboxylic acid
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only