missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropropane 99.0+%, TCI America™

Catalog No. C026625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C026625ML 25 mL
C0266500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C026625ML Supplier TCI America Supplier No. C026625ML

Chemical Identifiers

CAS 540-54-5
Molecular Formula C3H7Cl
Molecular Weight (g/mol) 78.539
MDL Number MFCD00000995
InChI Key SNMVRZFUUCLYTO-UHFFFAOYSA-N
Synonym propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
PubChem CID 10899
IUPAC Name 1-chloropropane
SMILES CCCCl

Specifications

Melting Point -123°C
Color Colorless
Boiling Point 46°C
Quantity 25 mL
UN Number 1278
Formula Weight 78.54
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloropropane
EINECSNumber (2)-0080
RTECSNumber TX4400000
TSCA Yes