missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropropane, 99%

Catalog No. AC109951000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109951000 100 mL Glass bottle
AC109955000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109951000 Supplier Thermo Scientific Chemicals Supplier No. 109951000

CAS: 540-54-5 | C3H7Cl | 78.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 540-54-5
Molecular Formula C3H7Cl
Molecular Weight (g/mol) 78.54
MDL Number MFCD00000995
InChI Key SNMVRZFUUCLYTO-UHFFFAOYSA-N
Synonym propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
PubChem CID 10899
IUPAC Name 1-chloropropane
SMILES CCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -123°C
Color Colorless
Density 0.8920g/mL
Boiling Point 46°C to 47°C
Flash Point -18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3870 to 1.389
Linear Formula CH3CH2CH2Cl
Quantity 100 mL
Beilstein 01, 104
Specific Gravity 0.892
Merck Index 15, 7960
Solubility Information Solubility in water: 2.7g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 78.54
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-chloropropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 208-749-7

RUO – Research Use Only