missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropinacolin 97.0+%, TCI America™

Catalog No. C133525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C133525ML 25 mL
C1335500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C133525ML Supplier TCI America Supplier No. C133525ML
Only null left

Chemical Identifiers

CAS 13547-70-1
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00035688
InChI Key ULSAJQMHTGKPIY-UHFFFAOYSA-N
Synonym 1-chloropinacolone, 1-chloro-3,3-dimethyl-2-butanone, chlorpinakolin, 2-butanone, 1-chloro-3,3-dimethyl, 1-monochloropinacoline, tert-butyl chloromethyl ketone, alpha-chloropinacolin, alpha-chloropinacoline, chloromethyl tert-butyl ketone, chloropinacoline
PubChem CID 83572
IUPAC Name 1-chloro-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CCl

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 mL
Formula Weight 134.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloropinacolin
EINECSNumber (2)-3557
TSCA Yes