accessibility menu, dialog, popup

UserName

CAS RN 13547-70-1

IUPAC Name: 1-chloro-3,3-dimethylbutan-2-one
Synonyms: 1-monochloropinacoline
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: ULSAJQMHTGKPIY-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results