missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorooctane 99.0+%, TCI America™
$41.02 - $106.25
Chemical Identifiers
| CAS | 111-85-3 |
|---|---|
| Molecular Formula | C8H17Cl |
| Molecular Weight (g/mol) | 148.674 |
| MDL Number | MFCD00001022 |
| InChI Key | CNDHHGUSRIZDSL-UHFFFAOYSA-N |
| Synonym | n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543 |
| PubChem CID | 8142 |
| IUPAC Name | 1-chlorooctane |
| SMILES | CCCCCCCCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C023625ML
|
TCI America
C023625ML |
25 mL |
Each for $41.02
|
|
|||||
|
C0236500ML
|
TCI America
C0236500ML |
500 mL |
Each for $106.25
|
|
|||||
Chemical Identifiers
| 111-85-3 | |
| 148.674 | |
| CNDHHGUSRIZDSL-UHFFFAOYSA-N | |
| 8142 | |
| CCCCCCCCCl |
| C8H17Cl | |
| MFCD00001022 | |
| n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543 | |
| 1-chlorooctane |
Specifications
| 111-85-3 | |
| 183°C | |
| MFCD00001022 | |
| 3082 | |
| CNDHHGUSRIZDSL-UHFFFAOYSA-N | |
| 1-chlorooctane | |
| 8142 | |
| ≥99.0% (GC) | |
| 1-Chlorooctane |
| Colorless | |
| C8H17Cl | |
| 25 mL | |
| n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543 | |
| CCCCCCCCCl | |
| 148.674 | |
| 148.67 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0066&(2)-0068
TSCA : Yes