accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 111-85-3

IUPAC Name: 1-chlorooctane
Synonyms: 1-chlorooctane
Molecular Weight (g/mol): 148.67
Molecular Formula: C8H17Cl
InChi Key: CNDHHGUSRIZDSL-UHFFFAOYSA-N
SMILES: CCCCCCCCCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)