missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctane 99.0+%, TCI America™

Catalog No. C023625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023625ML 25 mL
C0236500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C023625ML Supplier TCI America Supplier No. C023625ML

Chemical Identifiers

CAS 111-85-3
Molecular Formula C8H17Cl
Molecular Weight (g/mol) 148.674
MDL Number MFCD00001022
InChI Key CNDHHGUSRIZDSL-UHFFFAOYSA-N
Synonym n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543
PubChem CID 8142
IUPAC Name 1-chlorooctane
SMILES CCCCCCCCCl

Specifications

Color Colorless
Boiling Point 183°C
Quantity 25 mL
UN Number 3082
Formula Weight 148.67
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorooctane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes