Learn More
1-Chlorooctane, 99%
CAS: 111-85-3 | C8H17Cl | 148.674 g/mol
Supplier: Thermo Scientific Chemicals L04358AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Chlorooctane | |
| -58°C | |
| 182°C to 183°C | |
| C8H17Cl | |
| MFCD00001022 | |
| UN3082 | |
| n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543 | |
| CCCCCCCCCl | |
| 148.674 | |
| 148.68 |
| 111-85-3 | |
| 0.873 | |
| 63°C (145°F) | |
| 1.4298 | |
| 100 mL | |
| 1697464 | |
| CNDHHGUSRIZDSL-UHFFFAOYSA-N | |
| 1-chlorooctane | |
| 8142 | |
| 99% |
Chemical Identifiers
| 111-85-3 | |
| 148.674 | |
| CNDHHGUSRIZDSL-UHFFFAOYSA-N | |
| 8142 | |
| CCCCCCCCCl |
| C8H17Cl | |
| MFCD00001022 | |
| n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543 | |
| 1-chlorooctane |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 203-915-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only