missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctadecane 98.0+%, TCI America™

Catalog No. C023525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023525ML 25 mL
C0235500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C023525ML Supplier TCI America Supplier No. C023525ML

Chemical Identifiers

CAS 3386-33-2
Molecular Formula C18H37Cl
Molecular Weight (g/mol) 288.94
MDL Number MFCD00000960
InChI Key VUQPJRPDRDVQMN-UHFFFAOYSA-N
Synonym octadecyl chloride, octadecane, 1-chloro, n-octadecyl chloride, 1-chloroctadecane, chlorooctadecane, octadecane, chloro, unii-73h8vd533i, 1-chloranyloctadecane, stearylchloride, acmc-1cnf6
PubChem CID 18815
IUPAC Name 1-chlorooctadecane
SMILES CCCCCCCCCCCCCCCCCCCl

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 mL
Formula Weight 288.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorooctadecane
EINECSNumber (2)-0066&(2)-0068
RTECSNumber RG0187200
TSCA Yes
Recommended Storage Refrigerator