missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctadecane, 94%

Catalog No. AAB2490014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2490014 25 g
AAB2490022 100 g
AAB2490036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2490014 Supplier Thermo Scientific Chemicals Supplier No. B2490014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3386-33-2 | C18H37Cl | 288.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3386-33-2
Molecular Formula C18H37Cl
Molecular Weight (g/mol) 288.94
MDL Number MFCD00000960
InChI Key VUQPJRPDRDVQMN-UHFFFAOYSA-N
Synonym octadecyl chloride, octadecane, 1-chloro, n-octadecyl chloride, 1-chloroctadecane, chlorooctadecane, octadecane, chloro, unii-73h8vd533i, 1-chloranyloctadecane, stearylchloride, acmc-1cnf6
PubChem CID 18815
IUPAC Name 1-chlorooctadecane
SMILES CCCCCCCCCCCCCCCCCCCl

Specifications

Melting Point 20°C to 23°C
Density 0.849
Boiling Point 157°C to 158°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.451
Linear Formula CH3(CH2)17Cl
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 288.95
Percent Purity 94%
Chemical Name or Material 1-Chlorooctadecane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-207-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only