Learn More
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 98+%
CAS: 140681-55-6 | C7H14B2ClF9N2 | 354.26 g/mol
Supplier: Thermo Scientific Chemicals L1700314
Description
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) is used as a reagent for electrophilic fluorination and also acts as a selective oxidizing agent. It is a highly effective and versatile source of electrophilic fluorine and also used in the synthesis of organofluorine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) is used as a reagent for electrophilic fluorination and also acts as a selective oxidizing agent. It is a highly effective and versatile source of electrophilic fluorine and also used in the synthesis of organofluorine.
Solubility
Soluble in water, acetonitrile, alcohol, acetone, dimethylformamide, pyridine and dimethylsulfoxide.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases.
Specifications
| 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) | |
| ∽250°C (decomposition) | |
| MFCD00142607 | |
| UN3088 | |
| Soluble in water,acetonitrile,alcohol,acetone,dimethylformamide,pyridine and dimethylsulfoxide. | |
| F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 | |
| 354.26 | |
| 354.26 |
| 140681-55-6 | |
| C7H14B2ClF9N2 | |
| 25 g | |
| selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate | |
| TXRPHPUGYLSHCX-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate | |
| 2724933 | |
| ≥98% |
Chemical Identifiers
| 140681-55-6 | |
| 354.26 | |
| TXRPHPUGYLSHCX-UHFFFAOYSA-N | |
| 2724933 | |
| F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 |
| C7H14B2ClF9N2 | |
| MFCD00142607 | |
| selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate | |
| 1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate |
Safety and Handling
GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
H302-H317-H318
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: SELF-HEATING SOLID, ORGANIC, N.O.S.
EINECSNumber : 414-380-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only