missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Melting Point | 260.0°C |
| Color | White |
| Flash Point | >54°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (Iodometry) |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Merck Index | 15,8562 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in hcl,moderately soluble in ch3cn,dmf,slightly soluble in (ch3)2co |
| Formula Weight | 354.26 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. Harmful if swallowed. Harmful to aquatic life with long lasting effects. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protecti |
RUO – Research Use Only