missing translation for 'onlineSavingsMsg'
Learn More

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%

Catalog No. AC291490250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291490250 25 g Glass bottle
AC291490050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC291490250 Supplier Thermo Scientific Chemicals Supplier No. 291490250
Only null left

CAS: 140681-55-6 | C7H14B2ClF9N2 | 354.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 140681-55-6
Molecular Formula C7H14B2ClF9N2
Molecular Weight (g/mol) 354.26
MDL Number MFCD00142607
InChI Key TXRPHPUGYLSHCX-UHFFFAOYSA-N
Synonym selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate
PubChem CID 2724933
IUPAC Name 1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate
SMILES F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2

Specifications

Melting Point 260.0°C
Color White
Flash Point >54°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Iodometry)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,8562
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hcl,moderately soluble in ch3cn,dmf,slightly soluble in (ch3)2co
Formula Weight 354.26
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti

RUO – Research Use Only