missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexane 95.0+%, TCI America™

Catalog No. C018425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C018425ML 25 mL
C0184500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C018425ML Supplier TCI America Supplier No. C018425ML

Chemical Identifiers

CAS 544-10-5
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00001018
InChI Key MLRVZFYXUZQSRU-UHFFFAOYSA-N
Synonym chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride
PubChem CID 10992
IUPAC Name 1-chlorohexane
SMILES CCCCCCCl

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 mL
UN Number 1993
Formula Weight 120.62
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorohexane
EINECSNumber (2)-0066
TSCA Yes