missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorohexadecane 98.0+%, TCI America™
$50.33 - $164.03
Chemical Identifiers
| CAS | 4860-03-1 |
|---|---|
| Molecular Formula | C16H33Cl |
| Molecular Weight (g/mol) | 260.89 |
| MDL Number | MFCD00000959 |
| InChI Key | CLWAXFZCVYJLLM-UHFFFAOYSA-N |
| Synonym | cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580 |
| PubChem CID | 20993 |
| IUPAC Name | 1-chlorohexadecane |
| SMILES | CCCCCCCCCCCCCCCCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C018325ML
|
TCI America
C018325ML |
25 mL |
Each for $50.33
|
|
|||||
|
C0183500ML
|
TCI America
C0183500ML |
500 mL |
Each for $164.03
|
|
|||||
Chemical Identifiers
| 4860-03-1 | |
| 260.89 | |
| CLWAXFZCVYJLLM-UHFFFAOYSA-N | |
| 20993 | |
| CCCCCCCCCCCCCCCCCl |
| C16H33Cl | |
| MFCD00000959 | |
| cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580 | |
| 1-chlorohexadecane |
Specifications
| 4860-03-1 | |
| 195°C | |
| MFCD00000959 | |
| cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580 | |
| CCCCCCCCCCCCCCCCCl | |
| 260.89 | |
| 260.89 | |
| Liquid |
| Colorless | |
| C16H33Cl | |
| 25 mL | |
| CLWAXFZCVYJLLM-UHFFFAOYSA-N | |
| 1-chlorohexadecane | |
| 20993 | |
| ≥98.0% (GC) | |
| 1-Chlorohexadecane |
Safety and Handling
EINECSNumber : (2)-0066&(2)-0068
TSCA : Yes