missing translation for 'onlineSavingsMsg'
Learn More

(1-Chloroethyl)benzene 97.0+%, TCI America™

Catalog No. p-7124255
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C016625ML 25 mL
C0166500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C016625ML Supplier TCI America Supplier No. C016625ML
Only null left

Chemical Identifiers

CAS 672-65-1
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00018870
InChI Key GTLWADFFABIGAE-UHFFFAOYSA-N
Synonym 1-chloroethyl benzene, 1-chloro-1-phenylethane, 1-phenylethyl chloride, alpha-phenylethyl chloride, alpha-methylbenzyl chloride, benzene, 1-chloroethyl, 1-phenyl-ethylchloride, ccris 1778, .alpha.-methylbenzyl chloride, rs-phenethyl chloride
PubChem CID 12648
IUPAC Name 1-chloroethylbenzene
SMILES CC(C1=CC=CC=C1)Cl

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
UN Number 1993
Formula Weight 140.61
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Chloroethyl)benzene
EINECSNumber (3)-0033
TSCA Yes