accessibility menu, dialog, popup

UserName

H3CCl

CAS RN 672-65-1

IUPAC Name: (1-chloroethyl)benzene
Synonyms: (1-chloroethyl)benzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: GTLWADFFABIGAE-UHFFFAOYNA-N
SMILES: CC(Cl)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)