missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorododecane 97.0+%, TCI America™
$41.02 - $104.38
Chemical Identifiers
| CAS | 112-52-7 |
|---|---|
| Molecular Formula | C12H25Cl |
| Molecular Weight (g/mol) | 204.78 |
| MDL Number | MFCD00013688 |
| InChI Key | YAYNEUUHHLGGAH-UHFFFAOYSA-N |
| Synonym | lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk |
| PubChem CID | 8192 |
| IUPAC Name | 1-chlorododecane |
| SMILES | CCCCCCCCCCCCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C016325ML
|
TCI America
C016325ML |
25 mL |
Each for $41.02
|
|
|||||
|
C0163500ML
|
TCI America
C0163500ML |
500 mL |
Each for $104.38
|
|
|||||
Chemical Identifiers
| 112-52-7 | |
| 204.78 | |
| YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
| 8192 | |
| CCCCCCCCCCCCCl |
| C12H25Cl | |
| MFCD00013688 | |
| lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
| 1-chlorododecane |
Specifications
| 112-52-7 | |
| 260°C | |
| MFCD00013688 | |
| 3082 | |
| YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
| 1-chlorododecane | |
| 8192 | |
| ≥97.0% (GC) | |
| 1-Chlorododecane |
| Colorless | |
| C12H25Cl | |
| 25 mL | |
| lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
| CCCCCCCCCCCCCl | |
| 204.78 | |
| 204.78 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0066&(2)-0068
TSCA : Yes