missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Chloroacetyl)pyrrolidine 97.0+%, TCI America™
$90.59 - $451.29
Chemical Identifiers
| CAS | 20266-00-6 |
|---|---|
| Molecular Formula | C6H10ClNO |
| Molecular Weight (g/mol) | 147.602 |
| MDL Number | MFCD01217237 |
| InChI Key | AAOSLLBWWRKJIR-UHFFFAOYSA-N |
| PubChem CID | 88440 |
| IUPAC Name | 2-chloro-1-pyrrolidin-1-ylethanone |
| SMILES | C1CCN(C1)C(=O)CCl |
Chemical Identifiers
| 20266-00-6 | |
| 147.602 | |
| AAOSLLBWWRKJIR-UHFFFAOYSA-N | |
| 2-chloro-1-pyrrolidin-1-ylethanone |
| C6H10ClNO | |
| MFCD01217237 | |
| 88440 | |
| C1CCN(C1)C(=O)CCl |
Specifications
| 20266-00-6 | |
| White-Yellow | |
| C6H10ClNO | |
| 1 g | |
| C1CCN(C1)C(=O)CCl | |
| 147.602 | |
| 147.60 | |
| Crystalline Lumps |
| 46°C | |
| 105°C | |
| MFCD01217237 | |
| AAOSLLBWWRKJIR-UHFFFAOYSA-N | |
| 2-chloro-1-pyrrolidin-1-ylethanone | |
| 88440 | |
| ≥97.0% (GC) | |
| 1-(Chloroacetyl)pyrrolidine |
Safety and Handling
TSCA : No