accessibility menu, dialog, popup

UserName

CAS RN 20266-00-6

ClON

CAS RN 20266-00-6

IUPAC Name: 2-chloro-1-(pyrrolidin-1-yl)ethan-1-one
Synonyms: 2-chloro-1-(pyrrolidin-1-yl)ethanone
Molecular Weight (g/mol): 147.6
Molecular Formula: C6H10ClNO
InChi Key: AAOSLLBWWRKJIR-UHFFFAOYSA-N
SMILES: ClCC(=O)N1CCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results