missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloroacetyl)pyrrolidine 97.0+%, TCI America™

Catalog No. C29485G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29485G 5 g
C29481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29485G Supplier TCI America Supplier No. C29485G
Only null left

Chemical Identifiers

CAS 20266-00-6
Molecular Formula C6H10ClNO
Molecular Weight (g/mol) 147.602
MDL Number MFCD01217237
InChI Key AAOSLLBWWRKJIR-UHFFFAOYSA-N
PubChem CID 88440
IUPAC Name 2-chloro-1-pyrrolidin-1-ylethanone
SMILES C1CCN(C1)C(=O)CCl

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 147.60
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(Chloroacetyl)pyrrolidine