missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Chloroacetyl)pyrrolidine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 46°C |
| Color | White-Yellow |
| Boiling Point | 105°C |
| Quantity | 1 g |
| Formula Weight | 147.60 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 1-(Chloroacetyl)pyrrolidine |
Safety and Handling
| TSCA | No |