missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%

Catalog No. p-7036367
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54647MD 250 mg
AAH5464703 1 g
AAH5464706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54647MD Supplier Thermo Scientific Chemicals Supplier No. H54647MD
Only null left

CAS: 762240-99-3 | C4H4ClF3N2O2 | 204.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 762240-99-3
Molecular Formula C4H4ClF3N2O2
Molecular Weight (g/mol) 204.53
MDL Number MFCD07779449
InChI Key DYKIVKLXFDNBMY-UHFFFAOYSA-N
Synonym 1-chloroacetyl-2-trifluoroacetyl hydrazine, n'-2-chloroacetyl-2,2,2-trifluoroacetohydrazide, n'-chloroacetyl-2,2,2-trifluoroacetohydrazide, acetic acid, 2,2,2-trifluoro-, 2-2-chloroacetyl hydrazide, 2-chloro-n'-2,2,2-trifluoroacetyl acetohydrazide, n'-2-chloroacetyl-trifluoroacetohydrazide, 1-trifluoroacetyl-2-chloroacetyl hydrazine, 2-chloro-n'-trifluoroacetyl acetohydrazide, acetic acid, trifluoro-, 2-chloroacetyl hydrazide
PubChem CID 11961370
IUPAC Name N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide
SMILES FC(F)(F)C(=O)NNC(=O)CCl

Specifications

Quantity 250 mg
Formula Weight 204.53
Percent Purity 97%
Chemical Name or Material 1-Chloroacetyl-2-(trifluoroacetyl)hydrazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only