accessibility menu, dialog, popup

UserName

CAS RN 762240-99-3

IUPAC Name: 2-chloro-N'-(2,2,2-trifluoroacetyl)acetohydrazide
Synonyms: 2-chloro-N'-(2,2,2-trifluoroacetyl)acetohydrazide
Molecular Weight (g/mol): 204.53
Molecular Formula: C4H4ClF3N2O2
InChi Key: DYKIVKLXFDNBMY-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)NNC(=O)CCl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)