missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-iodobenzene 99.0+%, TCI America™

Catalog No. p-7124779
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C107525G 25 g
C1075250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C107525G Supplier TCI America Supplier No. C107525G

Chemical Identifiers

CAS 637-87-6
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.452
MDL Number MFCD00001053
InChI Key GWQSENYKCGJTRI-UHFFFAOYSA-N
Synonym 4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
PubChem CID 12510
IUPAC Name 1-chloro-4-iodobenzene
SMILES C1=CC(=CC=C1Cl)I

Specifications

Melting Point 56°C
Boiling Point 227°C
Quantity 25 g
Formula Weight 238.45
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-4-iodobenzene