missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-iodobenzene, 99%

CAS: 637-87-6 | C6H4ClI | 238.452 g/mol

Supplier:  Thermo Scientific Chemicals A1345936

 View more versions of this product

Catalog No. AAA1345936

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Chloro-4-iodobenzene
52°C to 56°C
108°C (226°F)
MFCD00001053
1634414
4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
C1=CC(=CC=C1Cl)I
238.452
238.46
637-87-6
226°C to 227°C
C6H4ClI
500 g
Light sensitive
GWQSENYKCGJTRI-UHFFFAOYSA-N
1-chloro-4-iodobenzene
12510
99%

Chemical Identifiers

637-87-6
238.452
GWQSENYKCGJTRI-UHFFFAOYSA-N
12510
C1=CC(=CC=C1Cl)I
C6H4ClI
MFCD00001053
4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
1-chloro-4-iodobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332-H335

EINECSNumber : 211-305-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only