missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-iodobenzene, 99%

Catalog No. AAA1345914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1345914 25 g
AAA1345922 100 g
AAA1345936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1345914 Supplier Thermo Scientific Chemicals Supplier No. A1345914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 637-87-6 | C6H4ClI | 238.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 637-87-6
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.452
MDL Number MFCD00001053
InChI Key GWQSENYKCGJTRI-UHFFFAOYSA-N
Synonym 4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
PubChem CID 12510
IUPAC Name 1-chloro-4-iodobenzene
SMILES C1=CC(=CC=C1Cl)I

Specifications

Melting Point 52°C to 56°C
Boiling Point 226°C to 227°C
Flash Point 108°C (226°F)
Quantity 25 g
Beilstein 1634414
Sensitivity Light sensitive
Formula Weight 238.46
Percent Purity 99%
Chemical Name or Material 1-Chloro-4-iodobenzene
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-305-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only