missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-4-ethynylbenzene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 47°C |
| Quantity | 1 g |
| Physical Form | Crystal-Powder at 20°C |
| UN Number | 1325 |
| Boiling Point | 60°C |
| Formula Weight | 136.58 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | 1-Chloro-4-ethynylbenzene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |