missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-ethylbenzene 98.0+%, TCI America™

Supplier:  TCI America C309125G

 View more versions of this product

Catalog No. C309125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Chloro-4-ethylbenzene
Yellow
25 g
C8H9Cl
1993
GPOFSFLJOIAMSA-UHFFFAOYSA-N
1-chloro-4-ethylbenzene
69330
≥98.0% (GC)
622-98-0
184°C
Clear Liquid at 20°C
MFCD00040855
benzene, 1-chloro-4-ethyl, p-chloroethylbenzene, 4-chloroethylbenzene, 4-ethylchlorobenzene, acmc-20aovw, p-chlorophenyl ethane, 4-chloro ethylbenzene, 1-ethyl-4-chlorobenzene, 4-chloro-1-ethylbenzene, 1-chloro-4-ethyl benzene
CCC1=CC=C(C=C1)Cl
140.61
140.61

Chemical Identifiers

622-98-0
140.61
GPOFSFLJOIAMSA-UHFFFAOYSA-N
69330
CCC1=CC=C(C=C1)Cl
C8H9Cl
MFCD00040855
benzene, 1-chloro-4-ethyl, p-chloroethylbenzene, 4-chloroethylbenzene, 4-ethylchlorobenzene, acmc-20aovw, p-chlorophenyl ethane, 4-chloro ethylbenzene, 1-ethyl-4-chlorobenzene, 4-chloro-1-ethylbenzene, 1-chloro-4-ethyl benzene
1-chloro-4-ethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions