missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-4-ethylbenzene, 97%, Thermo Scientific Chemicals
Description
1-Chloro-4-ethylbenzene is used in paint & coating industry, rubber industry, water-treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-4-ethylbenzene is used in paint &coating industry, rubber industry, water-treatment.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1-Chloro-4-ethylbenzene is used in paint &coating industry, rubber industry, water-treatment.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.03 |
| Boiling Point | 184°C to 185°C |
| Quantity | 5 g |
| Odor | Aromatic |
| Refractive Index | 1.5175 |
| Beilstein | 1099335 |
| Solubility Information | Difficult to mix. |
| Formula Weight | 140.61 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Chloro-4-ethylbenzene |
Safety and Handling
| Hazard Category | H227 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 210-763-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only