missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-nitrobenzene 99.0+%, TCI America™
$29.83 - $162.17
Chemical Identifiers
| CAS | 121-73-3 |
|---|---|
| Molecular Formula | C6H4ClNO2 |
| Molecular Weight (g/mol) | 157.55 |
| MDL Number | MFCD00007202 |
| InChI Key | KMAQZIILEGKYQZ-UHFFFAOYSA-N |
| Synonym | m-chloronitrobenzene, 3-nitrochlorobenzene, 3-chloronitrobenzene, m-nitrochlorobenzene, benzene, 1-chloro-3-nitro, chloro-m-nitrobenzene, 3-chloro-1-nitrobenzene, metachloronitrobenzene, 1-chloronitrobenzene, 1-nitro-3-chlorobenzene |
| PubChem CID | 8489 |
| ChEBI | CHEBI:82420 |
| IUPAC Name | 1-chloro-3-nitrobenzene |
| SMILES | [O-][N+](=O)C1=CC=CC(Cl)=C1 |
Chemical Identifiers
| 121-73-3 | |
| 157.55 | |
| KMAQZIILEGKYQZ-UHFFFAOYSA-N | |
| 8489 | |
| 1-chloro-3-nitrobenzene |
| C6H4ClNO2 | |
| MFCD00007202 | |
| m-chloronitrobenzene, 3-nitrochlorobenzene, 3-chloronitrobenzene, m-nitrochlorobenzene, benzene, 1-chloro-3-nitro, chloro-m-nitrobenzene, 3-chloro-1-nitrobenzene, metachloronitrobenzene, 1-chloronitrobenzene, 1-nitro-3-chlorobenzene | |
| CHEBI:82420 | |
| [O-][N+](=O)C1=CC=CC(Cl)=C1 |
Specifications
| 121-73-3 | |
| Yellow | |
| C6H4ClNO2 | |
| 25 g | |
| m-chloronitrobenzene, 3-nitrochlorobenzene, 3-chloronitrobenzene, m-nitrochlorobenzene, benzene, 1-chloro-3-nitro, chloro-m-nitrobenzene, 3-chloro-1-nitrobenzene, metachloronitrobenzene, 1-chloronitrobenzene, 1-nitro-3-chlorobenzene | |
| [O-][N+](=O)C1=CC=CC(Cl)=C1 | |
| 157.55 | |
| CHEBI:82420 | |
| ≥99.0% (GC) | |
| 1-Chloro-3-nitrobenzene |
| 46°C | |
| 236°C | |
| MFCD00007202 | |
| 1578 | |
| KMAQZIILEGKYQZ-UHFFFAOYSA-N | |
| 1-chloro-3-nitrobenzene | |
| 8489 | |
| 157.55 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0442
RTECSNumber : CZ0940000
TSCA : Yes