missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-nitrobenzene 99.0+%, TCI America™

Catalog No. C0221500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0221500G 500 g
C022125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0221500G Supplier TCI America Supplier No. C0221500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00007202
InChI Key KMAQZIILEGKYQZ-UHFFFAOYSA-N
Synonym m-chloronitrobenzene, 3-nitrochlorobenzene, 3-chloronitrobenzene, m-nitrochlorobenzene, benzene, 1-chloro-3-nitro, chloro-m-nitrobenzene, 3-chloro-1-nitrobenzene, metachloronitrobenzene, 1-chloronitrobenzene, 1-nitro-3-chlorobenzene
PubChem CID 8489
ChEBI CHEBI:82420
IUPAC Name 1-chloro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 46°C
Color Yellow
Boiling Point 236°C
Quantity 500 g
UN Number 1578
Formula Weight 157.55
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-3-nitrobenzene
EINECSNumber (3)-0442
RTECSNumber CZ0940000
TSCA Yes