missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-nitrobenzene 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 46°C |
| Color | Yellow |
| Boiling Point | 236°C |
| Quantity | 25 g |
| UN Number | 1578 |
| Formula Weight | 157.55 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Chloro-3-nitrobenzene |
Safety and Handling
| EINECSNumber | (3)-0442 |
| RTECSNumber | CZ0940000 |
| TSCA | Yes |