missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-methyl-2-butene (stabilized with K2CO3) 85.0+%, TCI America™

Catalog No. C129025ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C129025ML 25 mL
C1290250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C129025ML Supplier TCI America Supplier No. C129025ML

Chemical Identifiers

CAS 503-60-6
Molecular Formula C5H9Cl
Molecular Weight (g/mol) 104.577
MDL Number MFCD00000982
InChI Key JKXQKGNGJVZKFA-UHFFFAOYSA-N
Synonym 1-chloro-3-methyl-2-butene, prenyl chloride, 2-butene, 1-chloro-3-methyl, 3,3-dimethylallyl chloride, 3-methyl-2-butenyl chloride, 3-methylcrotyl chloride, unii-kmt7v9llbr, gamma,gamma-dimethylallyl chloride, kmt7v9llbr, .gamma.,.gamma.-dimethylallyl chloride
PubChem CID 10429
IUPAC Name 1-chloro-3-methylbut-2-ene
SMILES CC(=CCCl)C

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 mL
UN Number 1993
Formula Weight 104.58
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-methyl-2-butene (stabilized with K2CO3)
EINECSNumber (2)-3279
RTECSNumber EM4298500
TSCA Yes