accessibility menu, dialog, popup

UserName

CH3CH3Cl

CAS RN 503-60-6

IUPAC Name: 1-chloro-3-methylbut-2-ene
Synonyms: chloropren
Molecular Weight (g/mol): 104.58
Molecular Formula: C5H9Cl
InChi Key: JKXQKGNGJVZKFA-UHFFFAOYSA-N
SMILES: CC(C)=CCCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)