missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-methyl-2-butene, 95%, stab. with potassium carbonate

Catalog No. p-7023212
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1654509 10 g
AAA1654518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1654509 Supplier Thermo Scientific Chemicals Supplier No. A1654509
Only null left

CAS: 503-60-6 | C5H9Cl | 104.577 g/mol

1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.

Solubility
Miscible with chloroform, acetone, diethyl ether and alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.

Chemical Identifiers

CAS 503-60-6
Molecular Formula C5H9Cl
Molecular Weight (g/mol) 104.577
MDL Number MFCD00000982
InChI Key JKXQKGNGJVZKFA-UHFFFAOYSA-N
Synonym 1-chloro-3-methyl-2-butene, prenyl chloride, 2-butene, 1-chloro-3-methyl, 3,3-dimethylallyl chloride, 3-methyl-2-butenyl chloride, 3-methylcrotyl chloride, unii-kmt7v9llbr, gamma,gamma-dimethylallyl chloride, kmt7v9llbr, .gamma.,.gamma.-dimethylallyl chloride
PubChem CID 10429
IUPAC Name 1-chloro-3-methylbut-2-ene
SMILES CC(=CCCl)C

Specifications

Density 0.928
Boiling Point 107°C to 109°C
Flash Point 13°C (56°F)
Refractive Index 1.449
Linear Formula (CH3)2C=CHCH2Cl
Quantity 10 g
UN Number UN1993
Solubility Information Miscible with chloroform,acetone,diethyl ether and alcohol. Immiscible with water.
Formula Weight 104.57
Percent Purity 95%
Chemical Name or Material 1-Chloro-3-methyl-2-butene, stabilized with potassium carbonate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-972-7
RTECSNumber EM4298500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only