missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-iodopropane (stabilized with Copper chip) 98.0+%, TCI America™
$95.06 - $260.97
Chemical Identifiers
| CAS | 6940-76-7 |
|---|---|
| Molecular Formula | C3H6ClI |
| Molecular Weight (g/mol) | 204.435 |
| MDL Number | MFCD00001096 |
| InChI Key | SFOYQZYQTQDRIY-UHFFFAOYSA-N |
| Synonym | propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane |
| PubChem CID | 81363 |
| IUPAC Name | 1-chloro-3-iodopropane |
| SMILES | C(CCl)CI |
Chemical Identifiers
| 6940-76-7 | |
| 204.435 | |
| SFOYQZYQTQDRIY-UHFFFAOYSA-N | |
| 81363 | |
| C(CCl)CI |
| C3H6ClI | |
| MFCD00001096 | |
| propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane | |
| 1-chloro-3-iodopropane |
Specifications
| 6940-76-7 | |
| 171°C | |
| MFCD00001096 | |
| propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane | |
| C(CCl)CI | |
| 204.435 | |
| 204.44 | |
| Liquid |
| Yellow | |
| C3H6ClI | |
| 5 g | |
| SFOYQZYQTQDRIY-UHFFFAOYSA-N | |
| 1-chloro-3-iodopropane | |
| 81363 | |
| ≥98.0% (GC) | |
| 1-Chloro-3-iodopropane (stabilized with Copper chip) |
Safety and Handling
TSCA : Yes