Learn More
1-Chloro-3-iodopropane, 98%, stab. with copper
CAS: 6940-76-7 | C3H6ClI | 204.435 g/mol
Supplier: Thermo Scientific Chemicals A1348922
Description
1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.
Solubility
Difficult to mix.
Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 1-Chloro-3-iodopropane | |
| 6940-76-7 | |
| 170°C to 172°C | |
| C3H6ClI | |
| MFCD00001096 | |
| 1731115 | |
| propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane | |
| SFOYQZYQTQDRIY-UHFFFAOYSA-N | |
| 1-chloro-3-iodopropane | |
| 81363 | |
| 98% |
| stabilized with copper | |
| 1.904 | |
| >110°C (230°F) | |
| 1.5463 | |
| 100 g | |
| Light sensitive | |
| Difficult to mix. | |
| C(CCl)CI | |
| 204.435 | |
| 204.44 |
Chemical Identifiers
| 6940-76-7 | |
| 204.435 | |
| SFOYQZYQTQDRIY-UHFFFAOYSA-N | |
| 81363 | |
| C(CCl)CI |
| C3H6ClI | |
| MFCD00001096 | |
| propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane | |
| 1-chloro-3-iodopropane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-088-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only