missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodobenzene (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. p-7124269
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01905G 5 g
C019025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01905G Supplier TCI America Supplier No. C01905G
Only null left

Chemical Identifiers

CAS 625-99-0
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001046
InChI Key JMLWXCJXOYDXRN-UHFFFAOYSA-N
Synonym 3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
PubChem CID 12269
IUPAC Name 1-chloro-3-iodobenzene
SMILES ClC1=CC=CC(I)=C1

Specifications

Boiling Point 228°C
Quantity 5 g
Formula Weight 238.45
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-iodobenzene (stabilized with Copper chip)