missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-fluorobenzene 97.0+%, TCI America™

Catalog No. C064825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C064825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C064825G Supplier TCI America Supplier No. C064825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-98-9
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000569
InChI Key VZHJIJZEOCBKRA-UHFFFAOYSA-N
Synonym m-chlorofluorobenzene, 3-chlorofluorobenzene, benzene, 1-chloro-3-fluoro, m-fluorochlorobenzene, 3-fluorochlorobenzene, 1-fluoro-3-chlorobenzene, 1-chloro-3-fluoro-benzene, m-fluoro chlorobenzene, pubchem3419, 3-fluoro-chlorobenzene
PubChem CID 223087
IUPAC Name 1-chloro-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 g
UN Number 1993
Formula Weight 130.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-fluorobenzene