missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,5-dimethoxybenzene 97.0+%, TCI America™

Catalog No. C125810G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C125810G 10 g
C125825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C125810G Supplier TCI America Supplier No. C125810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7051-16-3
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00008382
InChI Key WQHNWJBSROXROL-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethoxybenzene, 3,5-dimethoxychlorobenzene, benzene, 1-chloro-3,5-dimethoxy, 5-chlororesorcinol dimethyl ether, 1,3-dimethoxy-5-chlorobenzene, pubchem3042, benzene,5-dimethoxy, acmc-209oev, wln: gr co1 eo1, 3,5-dimethoxyphenyl chloride
PubChem CID 81502
IUPAC Name 1-chloro-3,5-dimethoxybenzene
SMILES COC1=CC(=CC(=C1)Cl)OC

Specifications

Melting Point 35°C
Color White
Quantity 10 g
Physical Form Crystal-Lump at 20°C
Boiling Point 113°C
Formula Weight 172.61
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1-Chloro-3,5-dimethoxybenzene
RTECSNumber CZ0320000
TSCA No