accessibility menu, dialog, popup

UserName

CAS RN 7051-16-3

IUPAC Name: 1-chloro-3,5-dimethoxybenzene
Synonyms: 1-chloro-3,5-dimethoxybenzene
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: WQHNWJBSROXROL-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(Cl)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)