missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,4-dinitrobenzene, tech. 90%

Catalog No. p-7026660
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2277322 100 g
AAB2277330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2277322 Supplier Thermo Scientific Chemicals Supplier No. B2277322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-40-2 | C6H3ClN2O4 | 202.55 g/mol

1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 610-40-2
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00007212
InChI Key QVQSOXMXXFZAKU-UHFFFAOYSA-N
Synonym 1-chloro-3,4-dinitrobenzene, 3,4-dinitrochlorobenzene, benzene, 4-chloro-1,2-dinitro, unii-z29b1f274e, 1,2-dinitro-4-chlorobenzene, 1,2-dino2 4-cl benzene, 4-chlordinitrobenzol, acmc-209mog, 4-chloro-1,2-dinitrobenzen, ghl.pd_mitscher_leg0.928
PubChem CID 33097
IUPAC Name 4-chloro-1,2-dinitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point ∼20°C to 25°C
Density 1.687
Boiling Point 14°C to 16°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.587
Quantity 100 g
UN Number UN1577
Beilstein 521564
Solubility Information Insoluble in water.
Formula Weight 202.55
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Chloro-3,4-dinitrobenzene
Show More Show Less
Hazard Category H301+H311-H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORODINITROBENZENES, LIQUID.
EINECSNumber 210-223-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only