missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,4-difluorobenzene 98.0+%, TCI America™

Catalog No. C15995G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15995G Supplier TCI America Supplier No. C15995G

Specifications

CAS 696-02-6
Color Yellow
Boiling Point 127°C
Molecular Formula C6H3ClF2
Quantity 5 g
MDL Number MFCD00042572
UN Number 1993
Synonym 1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
InChI Key OPQMRQYYRSTBME-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)F)F
IUPAC Name 4-chloro-1,2-difluorobenzene
Molecular Weight (g/mol) 148.537
PubChem CID 136519
Formula Weight 148.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3,4-difluorobenzene
Show More Show Less