missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-octyne, 98%, Thermo Scientific Chemicals

$111.22 - $111.22

Chemical Identifiers

CAS 51575-83-8
Molecular Formula C8H13Cl
Molecular Weight (g/mol) 144.642
MDL Number MFCD00274223
InChI Key OUMUOQQKEAGFCJ-UHFFFAOYSA-N
Synonym 1-chloro-2-octyne, acmc-1asda, oumuoqqkeagfcj-uhfffaoysa
PubChem CID 4349444
IUPAC Name 1-chlorooct-2-yne
SMILES CCCCCC#CCCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5345206
View Documents
Thermo Scientific Chemicals
H5345206
5 g
Each for $111.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51575-83-8
144.642
OUMUOQQKEAGFCJ-UHFFFAOYSA-N
4349444
CCCCCC#CCCl
C8H13Cl
MFCD00274223
1-chloro-2-octyne, acmc-1asda, oumuoqqkeagfcj-uhfffaoysa
1-chlorooct-2-yne

Specifications

51575-83-8
81°C (15 mmHg)
C8H13Cl
1.46
5 g
OUMUOQQKEAGFCJ-UHFFFAOYSA-N
1-chlorooct-2-yne
4349444
98%
0.931
68°C (154°F)
CH3(CH2)4C≡CCH2Cl
MFCD00274223
1-chloro-2-octyne, acmc-1asda, oumuoqqkeagfcj-uhfffaoysa
CCCCCC#CCCl
144.642
144.64
1-Chloro-2-octyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only