accessibility menu, dialog, popup

UserName

CAS RN 51575-83-8

IUPAC Name: 1-chlorooct-2-yne
Synonyms: 1-chlorooct-2-yne
Molecular Weight (g/mol): 144.64
Molecular Formula: C8H13Cl
InChi Key: OUMUOQQKEAGFCJ-UHFFFAOYSA-N
SMILES: CCCCCC#CCCl
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)